Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109771.1 | 5 | 2312 | |||
GenBank mRNA | BC042427.1 | - | 5 | 2312 |