Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136398.1 | 1 | 375 | |||
GenBank mRNA | BI462408.1 | - | 1 | 338 |