Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027428.1 | 6 | 1912 | |||
NR_027429.1 | 5 | 1825 | ||||
NR_027430.1 | 5 | 1701 | ||||
NR_027431.2 | 5 | 1675 | ||||
NR_027432.1 | 4 | 1588 | ||||
GenBank mRNA | AK021673.1 | - | 5 | 1544 | ||
AK096758.1 | - | 6 | 1614 | |||
AK308811.1 | - | 4 | 1141 | |||
BC028046.1 | - | 4 | 1249 |