Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGEPL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102429.1 | 1 | 166 | |||
GenBank mRNA | BF695378.1 | - | 165 | |||
BX474356.1 | - | 1 | 243 | |||
DB142079.1 | - | 1 | 125 |
Gene Symbol: ANKAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510676.2 | XP_011508978.1 | 23 | 4849 |