Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101930071 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126041.1 | 2 | 256 | |||
GenBank mRNA | BU566442.1 | - | 2 | 213 |