Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110257.1 | 6 | 8392 | |||
GenBank mRNA | BX641130.1 | - | 6 | 8399 |