Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110621.1 | 2 | 294 | |||
GenBank mRNA | BC045746.2 | - | 1 | 48 | ||
BC067883.1 | - | 2 | 70 |