Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLJ27354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033981.1 | 1, 2 | 246 | |||
GenBank mRNA | AK130864.1 | - | 1, 2 | 246 |