Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC104968399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130143.1 | 3 | 1017 | |||
NR_130144.1 | 3 | 699 | ||||
NR_130145.1 | 4 | 824 | ||||
GenBank mRNA | BG201476.1 | - | 2 | 464 | ||
BG218162.1 | - | 2 | 473 | |||
BG721448.1 | - | 3 | 701 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG220191.1 | - | 2 | 461 | Chr. 18 - 3255436 - 3256322 [-] on Build GRCh38 |