Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPEAR-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026547.1 | 1 | 421 | |||
NR_026548.1 | 1 | 421 | ||||
GenBank mRNA | AF426269.1 | - | 1 | 421 | ||
AF426270.1 | - | 1 | 421 | |||
BC016690.2 | - | 1 | 19 | |||
BC064517.1 | - | 1 | 19 | |||
DA908807.1 | - | 1 | 16 |