Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125919.1 | 3 | 1119 | |||
GenBank mRNA | BC029598.1 | - | 3 | 1121 |