Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLJ36000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027084.1 | 2 | 3185 | |||
GenBank mRNA | AK093319.1 | - | 1 | 2122 | ||
BC039120.1 | - | 2 | 3187 |