Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLG-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103778.1 | 7 | 2511 | |||
NR_103779.1 | 3 | 1256 | ||||
GenBank mRNA | AK056431.1 | - | 7 | 2508 | ||
BC041408.1 | - | 3 | 1260 |