Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046649.1 | 1, 2 | 389 | |||
GenBank mRNA | BX104027.1 | - | 1, 2 | 389 |