Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LANCL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110604.1 | 2 | 182 | |||
NR_110605.1 | 2 | 182 | ||||
NR_110606.1 | 2 | 182 | ||||
GenBank mRNA | AL599033.1 | - | 2 | 152 | ||
BX119066.1 | - | 2 | 128 | |||
DA820426.1 | - | 2 | 182 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | F25413.1 | - | 2 | 156 | Chr. 2 - 210324744 - 210460516 [+] on Build GRCh38 |