Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LOC101927365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125843.1 | 2 | 344 | |||
GenBank mRNA | AK025621.1 | - | 1 | 363 | ||
BC029846.1 | - | 1 | 402 | |||
BC047485.1 | - | 1 | 402 | |||
DB459887.1 | - | 2 | 345 |
Gene Symbol: TSTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010141.1 | XP_016865630.1 | 1 | 470 |