Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SACS-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103450.1 | 2 | 1972 | |||
GenBank mRNA | AK057396.1 | - | 2 | 1972 | ||
DB525041.1 | - | 1 | 362 |