Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017013299.1 | XP_016868788.1 | 5 | 736 | ||
XM_017013300.1 | XP_016868789.1 | 4 | 767 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC022402.1 | - | 2 | 608 | Chr. 8 - 74052340 - 74099853 [+] on Build GRCh38 |