Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120474.1 | 5 | 1571 | |||
GenBank mRNA | AK093309.1 | - | 5 | 1537 |