Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APCDD1L-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034147.1 | 3, 4 | 616 | |||
GenBank mRNA | AK054637.1 | - | 3, 4 | 618 |