Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN16-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103455.1 | 3 | 1753 | |||
NR_103456.1 | 2 | 1556 | ||||
GenBank mRNA | AK127889.1 | - | 3 | 1731 | ||
BC043177.2 | - | 2 | 1106 |