Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF14-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036486.1 | 4 | 764 | |||
GenBank mRNA | AK093430.1 | - | 4 | 780 | ||
AK125748.1 | - | 4 | 764 |