Gene Symbol: LINC01024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102739.1 | 1 | 1581 | |||
NR_102740.1 | 2 | 944 | ||||
NR_102741.1 | 2 | 663 | ||||
GenBank mRNA | AL557445.3 | - | 2 | 396 | ||
BC042448.1 | - | 1 | 1212 | |||
BG772617.1 | - | 2 | 236 |