Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033979.2 | 3 | 460 | |||
NR_110458.1 | 3 | 734 | ||||
NR_110459.1 | 5 | 685 | ||||
NR_110460.1 | 5 | 655 | ||||
NR_110461.1 | 4 | 590 | ||||
NR_110462.1 | 2 | 571 | ||||
NR_110463.1 | 4 | 553 | ||||
NR_110464.1 | 4 | 541 | ||||
NR_110465.1 | 4 | 539 | ||||
NR_110466.1 | 3 | 469 | ||||
NR_110467.1 | 3 | 442 | ||||
NR_110468.1 | 3 | 430 | ||||
GenBank mRNA | BC030713.2 | - | 3 | 721 | ||
BC047481.1 | - | 3 | 721 | |||
CB153479.1 | - | 3 | 460 | |||
DA326484.1 | - | 5 | 645 | |||
DA398428.1 | - | 4 | 446 | |||
DA736032.1 | - | 4 | 591 | |||
DB045673.1 | - | 3 | 397 | |||
DB067012.1 | - | 3 | 426 | |||
DB100342.1 | - | 4 | 507 | |||
DB100521.1 | - | 4 | 454 |