Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024102.1 | 1 | 58 | |||
GenBank mRNA | BC105115.1 | - | 1 | 58 | ||
BC105117.1 | - | 1 | 58 | |||
BX116511.1 | - | 11 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA400173.1 | - | 11 | Chr. 21 - 33165535 - 33170189 [-] on Build GRCh38 |