Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGFLAM-AS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046219.1 | 4 | 1558 | |||
GenBank mRNA | BC014585.1 | - | 4 | 1562 |