Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3IP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034111.1 | 4 | 363 | |||
GenBank mRNA | BC031311.1 | - | 3 | 629 | ||
BX647114.1 | - | 4 | 319 |