Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRHDE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026837.1 | 2 | 3416 | |||
GenBank mRNA | AK095156.1 | - | 2 | 2433 | ||
CR936828.1 | - | 1 | 4199 |