Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTF3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034027.1 | 3 | 330 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI489969.1 | - | 3 | 298 | Chr. 11 - 33161688 - 33190248 [+] on Build GRCh38 |