Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF571-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038247.1 | 5 | 2341 | |||
NR_038248.1 | 6 | 2252 | ||||
NR_038249.1 | 4 | 2077 | ||||
NR_038250.1 | 3 | 2018 | ||||
GenBank mRNA | BC042488.1 | - | 5 | 2342 | ||
BC043284.1 | - | 6 | 2255 | |||
BC044917.1 | - | 4 | 2079 | |||
BC110871.1 | - | 4 | 1904 | |||
BX647800.1 | - | 3 | 2008 |