Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173687.3 | NP_775958.1 | 1 | 197 | ||
NR_126398.1 | 1 | 197 | ||||
NR_126399.1 | 1 | 197 | ||||
XM_017013344.1 | XP_016868833.1 | 1 | 423 | |||
GenBank mRNA | AK094450.1 | - | 1 | 145 | ||
BC063497.1 | - | 1 | 108 | |||
BC073830.1 | - | 1 | 145 | |||
BC120993.2 | - | 1 | 132 | |||
DB260703.1 | - | 1 | 197 |