Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053249.3 | NP_444479.2 | 2 | 658 | ||
GenBank mRNA | AY028606.1 | - | 2 | 666 | ||
BC108971.2 | - | 2 | 658 | 40047321 |