Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205361.1 | NP_001192290.1 | 6 | 797 | ||
NM_001205362.1 | NP_001192291.1 | 6 | 757 | |||
NM_033623.6 | NP_296372.3 | 6 | 744 | |||
GenBank mRNA | AF198092.1 | - | 6 | 635 | ||
AJ305034.1 | - | 6 | 797 | |||
AK008657.1 | - | 6 | 634 | |||
AK139845.1 | - | 6 | 747 | |||
AK155081.1 | - | 6 | 668 | |||
AK165651.1 | - | 6 | 670 | |||
AK165675.1 | - | 6 | 642 | |||
BC020161.1 | - | 6 | 720 | 3988680 | ||
BC031666.1 | - | 6 | 653 | 4012394 | ||
BC037431.1 | - | 6 | 653 | 2648141 |
Gene Symbol: Tes3-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029314.1 | - | 1 | 950 |