Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148935.2 | NP_683737.2 | 9 | 1049 | ||
GenBank mRNA | AF323488.1 | - | 9 | 1054 | ||
AK081735.1 | - | 7 | 754 | |||
AY039039.1 | - | 9 | 1095 | |||
BC092242.1 | - | 9 | 1038 | 30698162 | ||
BC130222.1 | - | 9 | 1050 | 40141600 |