Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054094.5 | NP_473435.1 | 9, 10 | 1266 | ||
GenBank mRNA | AB059428.1 | - | 9, 10 | 1270 | ||
AK149550.1 | - | 9, 10 | 1276 | |||
BC016414.1 | - | 9, 10 | 1263 | 4238964 | ||
BC022149.1 | - | 9, 10 | 1244 | 5102652 |