Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080444.4 | NP_536692.1 | 3 | 830 | ||
GenBank mRNA | AF398971.1 | - | 3 | 859 | ||
AK028538.1 | - | 3 | 830 | |||
AK052929.1 | - | 3 | 746 | |||
BC141311.1 | - | 3 | 821 | 9055923 | ||
BC141313.1 | - | 3 | 821 | 9055925 |