Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002581.4 | NP_002572.2 | 2, 3 | 1867 | ||
XM_017014784.1 | XP_016870273.1 | 2, 3 | 1907 | |||
GenBank mRNA | AK300128.1 | - | 1, 2 | 388 | ||
BC078657.1 | - | 2, 3 | 1497 | 30340336 | ||
X68280.1 | - | 2, 3 | 1258 |