Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025567.1 | NP_001020738.1 | 5 | 1111 | ||
NM_130865.3 | NP_570935.3 | 5 | 1126 | |||
GenBank mRNA | AB037698.1 | - | 5 | 1092 | ||
AF421857.1 | - | 4 | 1072 | |||
AF421858.1 | - | 4 | 1057 | |||
AF448118.1 | - | 5 | 1110 | |||
AF463513.1 | - | 4 | 1072 | |||
AF499446.1 | - | 4 | 2893 | |||
BC050912.1 | - | 5 | 1088 | 6417663 |