Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2j2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039157.2 | NP_001034246.1 | 8 | 886 | ||
NM_001039158.2 | NP_001034247.1 | 7 | 860 | |||
NM_001039159.2 | NP_001034248.1 | 7 | 723 | |||
NM_001284312.1 | NP_001271241.1 | 8 | 787 | |||
NM_001284314.1 | NP_001271243.1 | 6 | 642 | |||
NM_021402.6 | NP_067377.4 | 8 | 1023 | |||
GenBank mRNA | AF296656.1 | - | 6 | 543 | ||
AK004626.1 | - | 7 | 707 | |||
AK010302.1 | - | 8 | 831 | |||
AK076265.1 | - | 8 | 988 | |||
AW826087.1 | - | 7 | 544 | |||
BC065779.1 | - | 8 | 787 | 1429290 | ||
U93242.1 | - | 6 | 543 |