Gene Symbol: PARN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134477.2 | NP_001127949.1 | 22 | 1580 | ||
NM_001242992.1 | NP_001229921.1 | 21 | 1525 | |||
NM_002582.3 | NP_002573.1 | 22 | 1663 | |||
XM_011522510.2 | XP_011520812.1 | 22 | 1669 | |||
XM_011522511.2 | XP_011520813.1 | 22 | 1668 | |||
XM_011522513.2 | XP_011520815.1 | 22 | 1579 | |||
XM_017023258.1 | XP_016878747.1 | 21 | 1589 | |||
XM_017023259.1 | XP_016878748.1 | 18 | 1268 | |||
XM_017023260.1 | XP_016878749.1 | 16 | 1157 | |||
GenBank mRNA | AJ005698.1 | - | 22 | 1574 | ||
AK023866.1 | - | 15 | 948 | |||
AK293189.1 | - | 22 | 1572 | |||
AK299653.1 | - | 15 | 1105 | |||
AK301648.1 | - | 21 | 1515 | |||
AK315020.1 | - | 22 | 1659 | |||
BC044763.1 | - | 22 | 1601 | |||
BC050029.1 | - | 22 | 1601 | 4825218 |