Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080287.2 | NP_525026.2 | 20 | 2019 | ||
NM_207706.1 | NP_997589.1 | 21 | 2055 | |||
GenBank mRNA | AB093301.1 | - | 10 | 3290 | ||
AF398884.1 | - | 19 | 1918 | |||
AK004448.1 | - | 1 | 40 | |||
AK032033.1 | - | 21 | 2028 | |||
AK033918.1 | - | 20 | 2213 | |||
AK035710.1 | - | 5 | 405 | |||
AK038097.1 | - | 20 | 2008 | |||
AK038455.1 | - | 20 | 2125 | |||
AK045428.1 | - | 20 | 2020 | |||
AK047040.1 | - | 20 | 1981 | |||
AK050823.1 | - | 20 | 1982 | |||
AK053574.1 | - | 21 | 2019 | |||
AK129023.1 | - | 20 | 2019 | |||
AK205725.1 | - | 3 | 212 | |||
BC006678.1 | - | 6 | 610 | |||
BC023954.1 | - | 21 | 2040 | 5320429 | ||
BC033418.1 | - | 19 | 1949 |