Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021371.2 | NP_067346.1 | 4 | 1028 | ||
NM_181045.1 | NP_851388.1 | 5 | 728 | |||
GenBank mRNA | AB041539.1 | - | 4 | 1019 | ||
AF282251.1 | - | 5 | 710 | |||
AK035865.1 | - | 5 | 697 | |||
AK081011.1 | - | 4 | 462 | |||
AK158641.1 | - | 4 | 390 | |||
AK158892.1 | - | 4 | 1000 | |||
AK163648.1 | - | 4 | 491 | |||
BC057129.1 | - | 4 | 1007 | 6811387 | ||
BC058963.1 | - | 4 | 1007 | 6811387 |