Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_126166.4 | NP_569054.2 | 6 | 1602 | ||
GenBank mRNA | AF355152.1 | - | 4 | 1284 | ||
AF420279.1 | - | 3 | 1241 | |||
AK083977.1 | - | 6 | 1603 | |||
AK166177.1 | - | 5 | 1477 | |||
AK171438.1 | - | 6 | 1584 | |||
BC099937.1 | - | 6 | 1640 | 30084049 |