Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133202.2 | NP_573465.1 | 2 | 661 | ||
GenBank mRNA | AF283762.1 | - | 2 | 661 | ||
BC117988.2 | - | 2 | 640 | 40104412 | ||
BC117989.2 | - | 2 | 640 |
Set Membership: |
![]() |