Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133222.3 | NP_573485.2 | 13 | 1986 | ||
GenBank mRNA | AF385682.1 | - | 13 | 1989 | ||
BC017134.1 | - | 13 | 1921 | 4511572 |
Set Membership: |
![]() |