Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd209d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130904.2 | NP_570974.1 | 7 | 685 | ||
GenBank mRNA | AF373411.1 | - | 7 | 685 | ||
AF440280.1 | - | 6 | 591 | |||
AK136586.1 | - | 7 | 741 |