Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hook2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167991.1 | NP_001161463.1 | 3, 4 | 311 | ||
NM_133255.2 | NP_573518.2 | 3, 4 | 311 | |||
GenBank mRNA | AK030599.1 | - | 3, 4 | 304 | ||
AK042048.1 | - | 3, 4 | 299 | |||
AK136069.1 | - | 3, 4 | 305 | |||
AK145768.1 | - | 3, 4 | 279 | |||
AK161612.1 | - | 3, 4 | 264 | |||
AK166954.1 | - | 3, 4 | 291 | |||
AK169073.1 | - | 3, 4 | 279 | |||
BC019486.1 | - | 3, 4 | 287 | 4211990 | ||
BC029967.1 | - | 3, 4 | 252 | |||
BC055881.1 | - | 3, 4 | 257 | 3590336 | ||
BC080744.1 | - | 3, 4 | 301 | 30471369 | ||
CJ084759.1 | - | 3, 4 | 340 |