Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159321.1 | NP_001152793.1 | 3, 4 | 1021 | ||
NM_133355.1 | NP_579933.1 | 2, 3 | 325 | |||
GenBank mRNA | AF099933.2 | - | 2, 3 | 325 | ||
BC075624.1 | - | 377 | 6841572 | |||
DQ193535.1 | - | 3, 4 | 1021 |