Display Info New header:
tfpt-header-footer.tfcom-eks-na-comerce-gateway-prd.commerceprod.thermofisher.net cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@6f9216bf
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous enableSimpleHeaderValue:
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Special Offers
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search All
Popular Product Areas
TaqMan Assays
Gene Expression
Microbe & ABR Detection
Copy Number Variation
Mutation Detection
miRNA
SNP Genotyping
Primary Antibodies
Secondary Antibodies
Isotype Controls
Proteins & Peptides
ELISA Kits
All Documents & Support
Certificates
SDS
Manuals & Protocols
Product FAQs
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VMN1R18 SIRNA Assays ›
Gene Symbol:
Vmn1r18
Gene Name:
vomeronasal 1 receptor 18
Gene Aliases:
V1rc26
Chromosome Location:
Chr.6: 57389668 - 57390567 on Build GRCm38
UniGene ID:
Mm.377177
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s100940
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
40 nmol
InVivo | 5 nmol
20 nmol
40 nmol
250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
10 umol
250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
10 umol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
Displaying...
Build 38.1 Mouse Chr.6: s100940
×
Created with Raphaël 2.1.2
Vmn1r18
NM_13418
1.1
BC132061
.1
BC138409
.1
57,386,5
00
57,386,6
00
57,386,7
00
57,386,8
00
57,386,9
00
57,387,0
00
57,387,1
00
57,387,2
00
57,387,3
00
57,387,4
00
57,387,5
00
57,387,6
00
57,387,7
00
57,387,8
00
57,387,9
00
57,388,0
00
57,388,1
00
57,388,2
00
57,388,3
00
57,388,4
00
57,388,5
00
57,388,6
00
57,388,7
00
57,388,8
00
57,388,9
00
57,389,0
00
57,389,1
00
57,389,2
00
57,389,3
00
57,389,4
00
57,389,5
00
57,389,6
00
57,389,7
00
57,389,8
00
57,389,9
00
57,390,0
00
57,390,1
00
57,390,2
00
57,390,3
00
57,390,4
00
57,390,5
00
57,390,6
00
57,390,7
00
57,390,8
00
57,390,9
00
57,391,0
00
57,391,1
00
57,391,2
00
57,391,3
00
57,391,4
00
57,391,5
00
57,391,6
00
57,391,7
00
57,391,8
00
57,391,9
00
57,392,0
00
57,392,1
00
57,392,2
00
57,392,3
00
57,392,4
00
57,392,5
00
57,392,6
00
57,392,7
00
57,392,8
00
57,392,9
00
57,393,0
00
57,393,1
00
57,393,2
00
57,393,3
00
57,393,4
00
57,393,5
00
57,393,6
00
57,393,7
00
Show all RefSeq and GenBank
Back To Top
siRNA Details
Gene Symbol: Vmn1r18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_134181.1
NP_598942.1
1
334
GenBank mRNA
BC132061.1
-
1
433
40130338
BC138409.1
-
1
433
8861429
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
response to pheromone
pheromone binding
pheromone receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-59d6677d6-6dsvk:80/100.66.77.145:80.
git-commit: 1e3318a2800c8d0a8829a508293490082e5b9833
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.30.0-Offline