Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138660.2 | NP_619601.2 | 9 | 1678 | ||
GenBank mRNA | AF526276.1 | - | 9 | 1652 | ||
AJ292072.1 | - | 1892 | ||||
AK037399.1 | - | 3 | 273 | |||
AK139608.1 | - | 9 | 1717 | |||
AK145886.1 | - | 9 | 1452 | |||
BC034533.1 | - | 6 | 1322 | |||
BC060672.1 | - | 9 | 1506 | |||
BC141296.1 | - | 9 | 1791 | 9055908 |